Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-fluoroaniline, 98%

Click to view available options
Quantity:
5g
25g
1003-99-2
C6H5BrFN
190.015
MFCD00070750
FWTXFEKVHSFTDQ-UHFFFAOYSA-N
benzenamine, 2-bromo-5-fluoro,2-bromo-5-fluorobenzenamine,2-bromo-5-fluoro-aniline,2-bromo-5-fluoro-phenylamine,2-bromo-5-fluoro-benzenamine,2-bromo-5-fluorophenylamine,pubchem1524,5-fluoro-2-bromoaniline,2-bromo-5-fluoro aniline,acmc-1c3a5
2773317
2-bromo-5-fluoroaniline
C1=CC(=C(C=C1F)N)Br
1003-99-2
C6H5BrFN
190.015
MFCD00070750
FWTXFEKVHSFTDQ-UHFFFAOYSA-N
benzenamine, 2-bromo-5-fluoro,2-bromo-5-fluorobenzenamine,2-bromo-5-fluoro-aniline,2-bromo-5-fluoro-phenylamine,2-bromo-5-fluoro-benzenamine,2-bromo-5-fluorophenylamine,pubchem1524,5-fluoro-2-bromoaniline,2-bromo-5-fluoro aniline,acmc-1c3a5
2773317
2-bromo-5-fluoroaniline
C1=CC(=C(C=C1F)N)Br
Boiling Point 154°C (23mmHg)
Quantity 5g
Melting Point 42°C to 46°C
Flash Point >110°C (230°F)
UN Number UN2811
Formula Weight 190.01
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluoroaniline

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.