Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Bromo-5-fluorobenzoic acid, 98+%
Click to view available options
Quantity:
1g
5g
25g
Specifications
Quantity | 5g |
Melting Point | 154°C to 158°C |
Beilstein | 2575978 |
Formula Weight | 219.01 |
Percent Purity | ≥98% |
Chemical Name or Material | 2-Bromo-5-fluorobenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : Yes