Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-fluorobenzoic acid, 99%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
394-28-5
C7H4BrFO2
219.01
OQBMJMJZMDBQSM-UHFFFAOYSA-N
benzoic acid, 2-bromo-5-fluoro,5-fluoro-2-bromobenzoic acid,2-bromo-5-fluoro-benzoic acid,2-bromo-5-fluorobenzoicacid,2-bromo-5-fluorbenzoic acid,buttpark 19\01-66,pubchem1315,acmc-1ahhq,ksc226g3p,rarechem al bo 0747
2778181
2-bromo-5-fluorobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)Br
394-28-5
C7H4BrFO2
219.01
OQBMJMJZMDBQSM-UHFFFAOYSA-N
benzoic acid, 2-bromo-5-fluoro,5-fluoro-2-bromobenzoic acid,2-bromo-5-fluoro-benzoic acid,2-bromo-5-fluorobenzoicacid,2-bromo-5-fluorbenzoic acid,buttpark 19\01-66,pubchem1315,acmc-1ahhq,ksc226g3p,rarechem al bo 0747
2778181
2-bromo-5-fluorobenzoic acid
C1=CC(=C(C=C1F)C(=O)O)Br
Quantity 25g
Infrared Spectrum Authentic
Melting Point 156.0°C to 158.0°C
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 219.01
Percent Purity 99%
Chemical Name or Material 2-Bromo-5-fluorobenzoic acid

GHS H Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

GHS Signal Word: Warning