Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-fluorobenzonitrile, 98+%

Click to view available options
Quantity:
5g
25g
57381-39-2
C7H3BrFN
200.01
MFCD00142875
MDHNVHCZDCSTMS-UHFFFAOYSA-N
2-bromo-5-fluorobenzenecarbonitrile,2-bromo-5-fluoro-benzonitrile,benzonitrile, 2-bromo-5-fluoro,2-cyano-4-fluorobromobenzene,pubchem3218,bromofluorobenzonitrile2,acmc-1awgr,brc6h3 f cn,ksc496i7h,2-bromo-5-fluorobenzonitrile
93654
2-bromo-5-fluorobenzonitrile
C1=CC(=C(C=C1F)C#N)Br
57381-39-2
C7H3BrFN
200.01
MFCD00142875
MDHNVHCZDCSTMS-UHFFFAOYSA-N
2-bromo-5-fluorobenzenecarbonitrile,2-bromo-5-fluoro-benzonitrile,benzonitrile, 2-bromo-5-fluoro,2-cyano-4-fluorobromobenzene,pubchem3218,bromofluorobenzonitrile2,acmc-1awgr,brc6h3 f cn,ksc496i7h,2-bromo-5-fluorobenzonitrile
93654
2-bromo-5-fluorobenzonitrile
C1=CC(=C(C=C1F)C#N)Br
Applications
2-Bromo-5-fluorobenzonitrile is used in the synthesis of pyrimidine functional compounds acting as GABAAα2/α3 binding site agonists used in the treatment of anxiety disorders.

Solubility
Soluble in methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Quantity 5g
Melting Point 88°C to 92°C
UN Number UN3439
Beilstein 8678330
Formula Weight 200.01
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-fluorobenzonitrile

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.