Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-fluorobenzotrifluoride, 97%

Click to view available options
Quantity:
1g
5g
25g
40161-55-5
C7H3BrF4
242.999
MFCD00040937
AIDVAZGOACECLJ-UHFFFAOYSA-N
2-bromo-5-fluorobenzotrifluoride,1-bromo-4-fluoro-2-trifluoromethyl benzene,1-bromo-2-trifluoromethyl-4-fluorobenzene,4-fluoro-2-trifluoromethyl bromobenzene,2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene,benzene, 1-bromo-4-fluoro-2-trifluoromethyl,2-bromo-5-fluoro benzotrifluoride,pubchem1637,pubchem1643
520983
1-bromo-4-fluoro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1F)C(F)(F)F)Br
40161-55-5
C7H3BrF4
242.999
MFCD00040937
AIDVAZGOACECLJ-UHFFFAOYSA-N
2-bromo-5-fluorobenzotrifluoride,1-bromo-4-fluoro-2-trifluoromethyl benzene,1-bromo-2-trifluoromethyl-4-fluorobenzene,4-fluoro-2-trifluoromethyl bromobenzene,2-bromo-alpha,alpha,alpha,5-tetrafluorotoluene,benzene, 1-bromo-4-fluoro-2-trifluoromethyl,2-bromo-5-fluoro benzotrifluoride,pubchem1637,pubchem1643
520983
1-bromo-4-fluoro-2-(trifluoromethyl)benzene
C1=CC(=C(C=C1F)C(F)(F)F)Br
Boiling Point 136°C
Flash Point 63°C (145°F)
Quantity 25g
Beilstein 2643544
Formula Weight 243
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-fluorobenzotrifluoride

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning