Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-fluorocinnamic acid, 98%

Click to view available options
Quantity:
1g
5g
25g
202865-70-1
C9H6BrFO2
245.047
MFCD00142878
AWWFJLNOEZAVHZ-DAFODLJHSA-N
2-bromo-5-fluorocinnamic acid,3-2-bromo-5-fluorophenyl acrylic acid,e-3-2-bromo-5-fluorophenyl acrylic acid,3-2-bromo-5-fluoro-phenyl-acrylic acid,2e-3-2-bromo-5-fluorophenyl prop-2-enoic acid,3-2-bromo-5-fluorophenyl prop-2-enoic acid,rarechem al bk 0749,e-3-2-bromo-5-fluorophenyl acrylicacid,e-3-2-bromo-5-fluorophenyl prop-2-enoic acid,2e-3-2-bromo-5-fluorophenyl-2-propenoic acid
5708380
(E)-3-(2-bromo-5-fluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1F)C=CC(=O)O)Br
202865-70-1
C9H6BrFO2
245.047
MFCD00142878
AWWFJLNOEZAVHZ-DAFODLJHSA-N
2-bromo-5-fluorocinnamic acid,3-2-bromo-5-fluorophenyl acrylic acid,e-3-2-bromo-5-fluorophenyl acrylic acid,3-2-bromo-5-fluoro-phenyl-acrylic acid,2e-3-2-bromo-5-fluorophenyl prop-2-enoic acid,3-2-bromo-5-fluorophenyl prop-2-enoic acid,rarechem al bk 0749,e-3-2-bromo-5-fluorophenyl acrylicacid,e-3-2-bromo-5-fluorophenyl prop-2-enoic acid,2e-3-2-bromo-5-fluorophenyl-2-propenoic acid
5708380
(E)-3-(2-bromo-5-fluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1F)C=CC(=O)O)Br
Melting Point 188°C to 192°C
Quantity 25g
Formula Weight 245.05
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-fluorocinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning