Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-hydroxy-3-methylpyridine, 97%

Click to view available options
Quantity:
250mg
1g
1003711-43-0
C6H6BrNO
188.024
MFCD09839267
KCCYUVIIKCPVDH-UHFFFAOYSA-N
2-bromo-5-hydroxy-3-methylpyridine,2-bromo-5-hydroxy-3-picoline,pubchem5966,6-bromo-5-methyl-3-pyridinol,3-pyridinol,6-bromo-5-methyl,6-bromo-5-methyl-pyridin-3-ol,6-bromanyl-5-methyl-pyridin-3-ol,abbypharma ap-30-3129
29919363
6-bromo-5-methylpyridin-3-ol
CC1=CC(=CN=C1Br)O
1003711-43-0
C6H6BrNO
188.024
MFCD09839267
KCCYUVIIKCPVDH-UHFFFAOYSA-N
2-bromo-5-hydroxy-3-methylpyridine,2-bromo-5-hydroxy-3-picoline,pubchem5966,6-bromo-5-methyl-3-pyridinol,3-pyridinol,6-bromo-5-methyl,6-bromo-5-methyl-pyridin-3-ol,6-bromanyl-5-methyl-pyridin-3-ol,abbypharma ap-30-3129
29919363
6-bromo-5-methylpyridin-3-ol
CC1=CC(=CN=C1Br)O
Quantity 250mg
Color White
Formula Weight 188.02
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-hydroxy-3-methylpyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning