Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-hydroxybenzaldehyde, 95%

Click to view available options
Quantity:
5g
25g
100g
2973-80-0
C7H5BrO2
201.019
MFCD03033065
SCRQAWQJSSKCFN-UHFFFAOYSA-N
4-bromo-3-formylphenol,2-bromo-5-hydroxy-benzaldehyde,2-bromo-5-hydroxy benzaldehyde,benzaldehyde, 2-bromo-5-hydroxy,2-bromo-5-hydroxy benzaldehyde,2-bromo-5-hydroxybenzadehyde,benzaldehyde,2-bromo-5-hydroxy,pubchem14430,acmc-209haw,6-brom-3-oxy-benzaldehyd
387179
2-bromo-5-hydroxybenzaldehyde
C1=CC(=C(C=C1O)C=O)Br
2973-80-0
C7H5BrO2
201.019
MFCD03033065
SCRQAWQJSSKCFN-UHFFFAOYSA-N
4-bromo-3-formylphenol,2-bromo-5-hydroxy-benzaldehyde,2-bromo-5-hydroxy benzaldehyde,benzaldehyde, 2-bromo-5-hydroxy,2-bromo-5-hydroxy benzaldehyde,2-bromo-5-hydroxybenzadehyde,benzaldehyde,2-bromo-5-hydroxy,pubchem14430,acmc-209haw,6-brom-3-oxy-benzaldehyd
387179
2-bromo-5-hydroxybenzaldehyde
C1=CC(=C(C=C1O)C=O)Br
Applications
2-Bromo-5-hydroxybenzaldehyde is a useful reactant used in the synthesis of PDE4 inhibitors, BCL-XL and cancel cell growth inhibitors. Also an reactant used in the preparation of many pharmaceutical agents.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Melting Point 130°C to 135°C
Quantity 5g
UN Number UN3077
Beilstein 2079921
Formula Weight 201.02
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Bromo-5-hydroxybenzaldehyde

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.