Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-iodopyridine, 97%

Click to view available options
Quantity:
1g
5g
73290-22-9
C5H3BrIN
283.894
MFCD03095201
LLKRSJVPTKFSLS-UHFFFAOYSA-N
2-bromo-5-iodo-pyridine,pyridine, 2-bromo-5-iodo,5-iodo-2-bromopyridine,pubchem5969,5-iodo-2-bromo-pyridine,acmc-1bb8d,ksc493q0p,2-bromo-5-iodopyridine
4738271
2-bromo-5-iodopyridine
C1=CC(=NC=C1I)Br
73290-22-9
C5H3BrIN
283.894
MFCD03095201
LLKRSJVPTKFSLS-UHFFFAOYSA-N
2-bromo-5-iodo-pyridine,pyridine, 2-bromo-5-iodo,5-iodo-2-bromopyridine,pubchem5969,5-iodo-2-bromo-pyridine,acmc-1bb8d,ksc493q0p,2-bromo-5-iodopyridine
4738271
2-bromo-5-iodopyridine
C1=CC(=NC=C1I)Br
Sensitivity Light sensitive
Melting Point 123°C to 124°C
Quantity 1g
Beilstein 109100
Formula Weight 283.89
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-iodopyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures