Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-iodopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
73290-22-9
C5H3BrIN
283.89
LLKRSJVPTKFSLS-UHFFFAOYSA-N
2-bromo-5-iodo-pyridine,pyridine, 2-bromo-5-iodo,5-iodo-2-bromopyridine,pubchem5969,5-iodo-2-bromo-pyridine,acmc-1bb8d,ksc493q0p,2-bromo-5-iodopyridine
4738271
2-bromo-5-iodopyridine
C1=CC(=NC=C1I)Br
73290-22-9
C5H3BrIN
283.89
LLKRSJVPTKFSLS-UHFFFAOYSA-N
2-bromo-5-iodo-pyridine,pyridine, 2-bromo-5-iodo,5-iodo-2-bromopyridine,pubchem5969,5-iodo-2-bromo-pyridine,acmc-1bb8d,ksc493q0p,2-bromo-5-iodopyridine
4738271
2-bromo-5-iodopyridine
C1=CC(=NC=C1I)Br
Packaging Glass Bottle
Melting Point 123°C to 123.6°C
Infrared Spectrum Authentic
Color White
Quantity 25g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 283.89
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-iodopyridine
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/e

GHS Signal Word: Warning