Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-methoxybenzyl bromide, 97%

Click to view available options
Quantity:
5g
25g
19614-12-1
C8H8Br2O
279.959
MFCD00078664
MURVUTUZSUEIGI-UHFFFAOYSA-N
2-bromo-5-methoxybenzyl bromide,1-bromo-2-bromomethyl-4-methoxybenzene,2-bromo-5-methoxybenzylbromide,4-bromo-3-bromomethyl anisole,2-bromo-5-methoxy-benzyl bromide,benzene, 1-bromo-2-bromomethyl-4-methoxy,4-bromo-3-bromomethyl-1-methoxybenzene,pubchem18176,acmc-20a5x2,4-bromo3-bromomethylanisole
3525951
1-bromo-2-(bromomethyl)-4-methoxybenzene
COC1=CC(=C(C=C1)Br)CBr
19614-12-1
C8H8Br2O
279.959
MFCD00078664
MURVUTUZSUEIGI-UHFFFAOYSA-N
2-bromo-5-methoxybenzyl bromide,1-bromo-2-bromomethyl-4-methoxybenzene,2-bromo-5-methoxybenzylbromide,4-bromo-3-bromomethyl anisole,2-bromo-5-methoxy-benzyl bromide,benzene, 1-bromo-2-bromomethyl-4-methoxy,4-bromo-3-bromomethyl-1-methoxybenzene,pubchem18176,acmc-20a5x2,4-bromo3-bromomethylanisole
3525951
1-bromo-2-(bromomethyl)-4-methoxybenzene
COC1=CC(=C(C=C1)Br)CBr
Applications
Employed as an important intermediate for raw material for organic synthesis, agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in hot methanol. Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 25g
Color White
Melting Point 90°C to 94°C (literature)
UN Number UN3261
Beilstein 1945118
Formula Weight 279.96
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-methoxybenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.