Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-methylpyridine, 98+%

Click to view available options
Quantity:
5g
25g
100g
3510-66-5
C6H6BrN
172.025
MFCD00209553
YWNJQQNBJQUKME-UHFFFAOYSA-N
2-bromo-5-picoline,6-bromo-3-picoline,2-bromo-5-methyl-pyridine,pyridine, 2-bromo-5-methyl,2-bromo-5-methyl pyridine,5-methyl-2-bromopyridine,2-brom-5-methylpyridin,pubchem1185
564216
2-bromo-5-methylpyridine
CC1=CN=C(C=C1)Br
3510-66-5
C6H6BrN
172.025
MFCD00209553
YWNJQQNBJQUKME-UHFFFAOYSA-N
2-bromo-5-picoline,6-bromo-3-picoline,2-bromo-5-methyl-pyridine,pyridine, 2-bromo-5-methyl,2-bromo-5-methyl pyridine,5-methyl-2-bromopyridine,2-brom-5-methylpyridin,pubchem1185
564216
2-bromo-5-methylpyridine
CC1=CN=C(C=C1)Br
Boiling Point 95°C to 96°C (12mmHg)
Melting Point 40°C to 45°C
Flash Point 103°C (217°F)
Quantity 100g
Beilstein 107323
Formula Weight 172.03
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-5-methylpyridine

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger