Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-methylpyrimidine, 95%

Click to view available options
Quantity:
250mg
1g
150010-20-1
C5H5BrN2
173.013
MFCD09753737
KYCGEJNZMHUBMX-UHFFFAOYSA-N
pyrimidine, 2-bromo-5-methyl,acmc-20a0od,2-bromo-5-methyl-pyrimidine,2-?bromo-?5-?methylpyrimidine
45789625
2-bromo-5-methylpyrimidine
CC1=CN=C(N=C1)Br
150010-20-1
C5H5BrN2
173.013
MFCD09753737
KYCGEJNZMHUBMX-UHFFFAOYSA-N
pyrimidine, 2-bromo-5-methyl,acmc-20a0od,2-bromo-5-methyl-pyrimidine,2-?bromo-?5-?methylpyrimidine
45789625
2-bromo-5-methylpyrimidine
CC1=CN=C(N=C1)Br
Quantity 250mg
Beilstein 21248671
Formula Weight 173.01
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 2-Bromo-5-methylpyrimidine