Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-nitroaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
10403-47-1
C6H5BrN2O2
217.02
MFCD00051578
BAAUCXCLMDAZEL-UHFFFAOYSA-N
benzenamine, 2-bromo-5-nitro,2-bromo-5-nitrobenzenamine,3-amino-4-bromonitrobenzene,2-bromo-5-nitro-phenylamine,2-bromo-5-nitrophenylamine,pubchem21903,2-bromo 5-nitroaniline,2-bromo-5-nitro-aniline,2-bromo-5-nitro aniline,acmc-2098ai
82607
2-bromo-5-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)Br
10403-47-1
C6H5BrN2O2
217.02
MFCD00051578
BAAUCXCLMDAZEL-UHFFFAOYSA-N
benzenamine, 2-bromo-5-nitro,2-bromo-5-nitrobenzenamine,3-amino-4-bromonitrobenzene,2-bromo-5-nitro-phenylamine,2-bromo-5-nitrophenylamine,pubchem21903,2-bromo 5-nitroaniline,2-bromo-5-nitro-aniline,2-bromo-5-nitro aniline,acmc-2098ai
82607
2-bromo-5-nitroaniline
C1=CC(=C(C=C1[N+](=O)[O-])N)Br
Melting Point 138°C to 142°C
Infrared Spectrum Authentic
Quantity 25g
Color Khaki to Yellow
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 217.02
Physical Form Fine Crystalline Needles or Crystalline Powder
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitroaniline
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-874-9