Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-nitroanisole, 98%

Click to view available options
Quantity:
2g
10g
77337-82-7
C7H6BrNO3
232.033
MFCD00041250
NTKADLOYTKVXQN-UHFFFAOYSA-N
2-bromo-5-nitroanisole,4-bromo-3-methoxynitrobenzene,2-bromo-5-nitroanisol,benzene, 1-bromo-2-methoxy-4-nitro,1-bromo-2-methoxy-4-nitro-benzene,zlchem 673,pubchem2536,2-bromo 5-nitro anisole,2-bromo-5-nitro-anisole,2-bromo-5-nitro anisole
101293
1-bromo-2-methoxy-4-nitrobenzene
COC1=C(C=CC(=C1)[N+](=O)[O-])Br
77337-82-7
C7H6BrNO3
232.033
MFCD00041250
NTKADLOYTKVXQN-UHFFFAOYSA-N
2-bromo-5-nitroanisole,4-bromo-3-methoxynitrobenzene,2-bromo-5-nitroanisol,benzene, 1-bromo-2-methoxy-4-nitro,1-bromo-2-methoxy-4-nitro-benzene,zlchem 673,pubchem2536,2-bromo 5-nitro anisole,2-bromo-5-nitro-anisole,2-bromo-5-nitro anisole
101293
1-bromo-2-methoxy-4-nitrobenzene
COC1=C(C=CC(=C1)[N+](=O)[O-])Br
Quantity 2g
Melting Point 102°C to 107°C
Beilstein 2520496
Formula Weight 232.03
Percent Purity 98%
Chemical Name or Material 2-Bromo-5-nitroanisole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning