Alfa Aesar™ 2-Bromo-5-nitrobenzotrifluoride, 97%

Manufacturer: Alfa Aesar™ A1141614

Quantity 25g

 View more versions of this product

Catalog No. AAA1141614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 367-67-9
    Molecular Formula C7H3BrF3NO2
    Formula Weight 270.01
    MDL Number MFCD00014707
    Beilstein 2460260
    Synonym 1-Bromo-4-nitro-2-(trifluoromethyl)benzene
    Chemical Name or Material 2-Bromo-5-nitrobenzotrifluoride

    Specifications

    Formula Weight 270.01
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    Quantity 25g
    Flash Point >110°C (230°F)
    Health Hazard 1 Warning
    CAS 367-67-9
    Boiling Point 87° to 88°C (3mmHg)
    Molecular Formula C7H3BrF3NO2
    MDL Number MFCD00014707
    Melting Point 41° to 45°C
    Beilstein 2460260
    Synonym 1-Bromo-4-nitro-2-(trifluoromethyl)benzene
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 2-Bromo-5-nitrobenzotrifluoride
    Assay 97%

    GHS H Statement
    H302-H312-H332-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280h-P305+P351+P338-P337+P313
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit