Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-5-nitrothiophene, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13195-50-1
C4H2BrNO2S
208.03
ZPNFMDYBAQDFDY-UHFFFAOYSA-N
5-bromo-2-nitrothiophene,thiophene, 2-bromo-5-nitro,pubchem5283,acmc-1bsij,2-nitro-5-bromothiophene,5-nitro-2-bromothiophene,5-bromo-2-nitro-thiophene,2-bromo-5-nitro-thiophene,labotest-bb lt00455100,thiophene,2-bromo-5-nitro
83222
2-bromo-5-nitrothiophene
C1=C(SC(=C1)Br)[N+](=O)[O-]
13195-50-1
C4H2BrNO2S
208.03
ZPNFMDYBAQDFDY-UHFFFAOYSA-N
5-bromo-2-nitrothiophene,thiophene, 2-bromo-5-nitro,pubchem5283,acmc-1bsij,2-nitro-5-bromothiophene,5-nitro-2-bromothiophene,5-bromo-2-nitro-thiophene,2-bromo-5-nitro-thiophene,labotest-bb lt00455100,thiophene,2-bromo-5-nitro
83222
2-bromo-5-nitrothiophene
C1=C(SC(=C1)Br)[N+](=O)[O-]
Melting Point 44°C to 48°C
Packaging Glass bottle
Quantity 25g
Assay Percent Range 96% min. (GC)
Formula Weight 208.03
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-nitrothiophene

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 236-155-8