Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-(trifluoromethyl)benzoic acid, 97%

Click to view available options
Quantity:
5g
25g
1483-56-3
C8H4BrF3O2
269.017
MFCD06208253
REBQGRPKXYIJDC-UHFFFAOYSA-N
2-bromo-5-trifluoromethyl benzoic acid,2-bromo-5-trifluoromethyl-benzoic acid,benzoic acid, 2-bromo-5-trifluoromethyl,pubchem4736,acmc-209d0f,ksc495e1n,rarechem al bo 1009,2-bromo-5-trifluoro methyl benzoic acid,2-bromo-5-trifluormethyl benzoic acid
15261013
2-bromo-5-(trifluoromethyl)benzoic acid
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Br
1483-56-3
C8H4BrF3O2
269.017
MFCD06208253
REBQGRPKXYIJDC-UHFFFAOYSA-N
2-bromo-5-trifluoromethyl benzoic acid,2-bromo-5-trifluoromethyl-benzoic acid,benzoic acid, 2-bromo-5-trifluoromethyl,pubchem4736,acmc-209d0f,ksc495e1n,rarechem al bo 1009,2-bromo-5-trifluoro methyl benzoic acid,2-bromo-5-trifluormethyl benzoic acid
15261013
2-bromo-5-(trifluoromethyl)benzoic acid
C1=CC(=C(C=C1C(F)(F)F)C(=O)O)Br
Applications
Used as a intermediate for pharmaceutical and precursor for organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 25g
Formula Weight 269.02
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Bromo-5-(trifluoromethyl)benzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning