Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-5-(trifluoromethyl)pyridine, 96%

Click to view available options
Quantity:
0.25g
1g
5g
50488-42-1
C6H3BrF3N
225.996
MFCD00153086
GSKMWMFOQQBVMI-UHFFFAOYSA-N
2-bromo-5-trifluoromethyl pyridine,6-bromo-3-trifluoromethylpyridine,2-bromo-5-trifluoromethyl-pyridine,pyridine, 2-bromo-5-trifluoromethyl,5-trifluoromethyl-2-bromopyridine,2-brom-5-trifluormethyl pyridin,2-bromo-5-trilfuoromethylpyridine,pubchem3005
2736434
2-bromo-5-(trifluoromethyl)pyridine
C1=CC(=NC=C1C(F)(F)F)Br
50488-42-1
C6H3BrF3N
225.996
MFCD00153086
GSKMWMFOQQBVMI-UHFFFAOYSA-N
2-bromo-5-trifluoromethyl pyridine,6-bromo-3-trifluoromethylpyridine,2-bromo-5-trifluoromethyl-pyridine,pyridine, 2-bromo-5-trifluoromethyl,5-trifluoromethyl-2-bromopyridine,2-brom-5-trifluormethyl pyridin,2-bromo-5-trilfuoromethylpyridine,pubchem3005
2736434
2-bromo-5-(trifluoromethyl)pyridine
C1=CC(=NC=C1C(F)(F)F)Br
Melting Point 45°C to 47°C
Quantity 0.25g
Boiling Point 78°C to 81°C (30mmHg)
Solubility Information Soluble in methanol and ethanol.
Formula Weight 226
Physical Form Crystalline
Percent Purity 96%
Chemical Name or Material 2-Bromo-5- (trifluoromethyl)pyridine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures