Alfa Aesar™ 2-Bromo-6-chloro-4-fluorophenol, 97%

Manufacturer: Alfa Aesar™ H2673403

Quantity 1g

 View more versions of this product

Catalog No. AAH2673403

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 886499-83-8
    Molecular Formula C6H3BrClFO
    Formula Weight 225.45
    MDL Number MFCD04115954
    Chemical Name or Material 2-Bromo-6-chloro-4-fluorophenol

    Specifications

    Formula Weight 225.45
    Molecular Formula C6H3BrClFO
    MDL Number MFCD04115954
    Melting Point 38° to 40°C
    Quantity 1g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 886499-83-8
    Chemical Name or Material 2-Bromo-6-chloro-4-fluorophenol
    Assay 97%

    Description

    Applications
    2-Bromo-6-chloro-4-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H302-H315-H319-H335
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit