Alfa Aesar™ 2-Bromo-6-chlorotoluene, 99%

Manufacturer: Alfa Aesar™ B2569714

Quantity 25g

 View more versions of this product

Catalog No. AAB2569714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62356-27-8
    Molecular Formula C7H6BrCl
    Formula Weight 205.48
    MDL Number MFCD00061121
    Chemical Name or Material 2-Bromo-6-chlorotoluene

    Specifications

    Formula Weight 205.48
    Boiling Point 120°C (40mmHg)
    Quantity 25g
    Flash Point 95°C (203°F)
    Molecular Formula C7H6BrCl
    MDL Number MFCD00061121
    CAS 62356-27-8
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.
    Chemical Name or Material 2-Bromo-6-chlorotoluene
    Assay 99%

    GHS H Statement
    H315-H319
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Specific treatment (see label).
    If skin irritation occurs: Get medical advice/attention.
    If eye irritation persists: Get medical advice/attention.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit