Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-6-fluorobenzaldehyde, 98%

Click to view available options
Quantity:
5g
25g
100g
360575-28-6
C7H4BrFO
203.01
MFCD03407341
PJNILWKRAKKEQM-UHFFFAOYSA-N
6-bromo-2-fluorobenzaldehyde,2-bromo-6-fluorobenzenecarbaldehyde,2-bromo-6-fluoro-benzaldehyde,2-fluoro-6-bromobenzaldehyde,6-fluoro-2-bromobenzaldehyde,benzaldehyde, 2-bromo-6-fluoro,pubchem1419,acmc-1afpf,ksc497k8t
22473977
2-bromo-6-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)C=O)F
360575-28-6
C7H4BrFO
203.01
MFCD03407341
PJNILWKRAKKEQM-UHFFFAOYSA-N
6-bromo-2-fluorobenzaldehyde,2-bromo-6-fluorobenzenecarbaldehyde,2-bromo-6-fluoro-benzaldehyde,2-fluoro-6-bromobenzaldehyde,6-fluoro-2-bromobenzaldehyde,benzaldehyde, 2-bromo-6-fluoro,pubchem1419,acmc-1afpf,ksc497k8t
22473977
2-bromo-6-fluorobenzaldehyde
C1=CC(=C(C(=C1)Br)C=O)F
Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
Density 1.70g/mL
Boiling Point 100°C to 102°C (8mmHg)
Quantity 100g
Melting Point 43°C to 47°C
Sensitivity Light Sensitive
Beilstein 2573976
Formula Weight 203.01
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-fluorobenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C