Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-6-hydrazinopyridine, 98%

Click to view available options
Quantity:
1g
5g
25g
26944-71-8
C5H6BrN3
188.028
MFCD09743754
PQMFVUNERGGBPG-UHFFFAOYSA-N
2-bromo-6-hydrazinylpyridine,2-bromo-6-hydrazinopyridine,2-6-bromopyridin-2-yl hydrazine,6-bromo-pyridin-2-yl-hydrazine,2-bromo-6-hydrazinopydine,6-bromo-2-pyridyl hydrazine,6-bromopyridin-2-yl hydrazine,6-bromo-2-hydrazino pyridine,2 1h-pyridinone, 6-bromo-, hydrazone,pubchem12987
19392588
(6-bromopyridin-2-yl)hydrazine
C1=CC(=NC(=C1)Br)NN
26944-71-8
C5H6BrN3
188.028
MFCD09743754
PQMFVUNERGGBPG-UHFFFAOYSA-N
2-bromo-6-hydrazinylpyridine,2-bromo-6-hydrazinopyridine,2-6-bromopyridin-2-yl hydrazine,6-bromo-pyridin-2-yl-hydrazine,2-bromo-6-hydrazinopydine,6-bromo-2-pyridyl hydrazine,6-bromopyridin-2-yl hydrazine,6-bromo-2-hydrazino pyridine,2 1h-pyridinone, 6-bromo-, hydrazone,pubchem12987
19392588
(6-bromopyridin-2-yl)hydrazine
C1=CC(=NC(=C1)Br)NN
Color Brown
Quantity 1g
Melting Point 117°C to 118°C
UN Number UN2811
Beilstein 386966
Formula Weight 188.03
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-hydrazinopyridine

GHS H Statement
H301-H318-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.