Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-(hydroxymethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog Number ac426310010
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
33674-96-3
C6H6BrNO
188.02
MFCD04039884
XDDGKNRSCDEWBR-UHFFFAOYSA-N
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
3517811
(6-bromopyridin-2-yl)methanol
OCC1=CC=CC(Br)=N1
33674-96-3
C6H6BrNO
188.02
MFCD04039884
XDDGKNRSCDEWBR-UHFFFAOYSA-N
6-bromopyridin-2-yl methanol, 2-bromo-6-pyridinemethanol, 2-bromo-6-hydroxymethyl pyridine, 6-bromo-2-hydroxymethyl pyridine, 2-pyridinemethanol, 6-bromo, 6-bromopyridine-2-methanol, 6-bromo-pyridine-2-yl methanol, 6-bromo-2-pyridinemethanol, 6-bromo-pyridin-2-yl-methanol, 6-bromo-pyridin-2-yl methanol
3517811
(6-bromopyridin-2-yl)methanol
OCC1=CC=CC(Br)=N1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 95.0
CAS Max % 100.0
Melting Point 34°C to 39°C
Color White
Boiling Point 246°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 188.02
Percent Purity 96%
Physical Form Powder
Chemical Name or Material 2-Bromo-6-(hydroxymethyl)pyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit