Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-6-methoxyphenol, 98+%

Click to view available options
Quantity:
1g
5g
25g
28165-49-3
C7H7BrO2
203.035
MFCD08146628
WEUFQISIJPSTBM-UHFFFAOYSA-N
2-bromo-6-methoxy-phenol,phenol, 2-bromo-6-methoxy,bromhydroxyanisol,6-bromoguaiacol,3-bromo-2-hydroxyanisole,acmc-1ch0f,phenol,2-bromo-6-methoxy,2-bromanyl-6-methoxy-phenol,ksc560s4p
11019958
2-bromo-6-methoxyphenol
COC1=C(C(=CC=C1)Br)O
28165-49-3
C7H7BrO2
203.035
MFCD08146628
WEUFQISIJPSTBM-UHFFFAOYSA-N
2-bromo-6-methoxy-phenol,phenol, 2-bromo-6-methoxy,bromhydroxyanisol,6-bromoguaiacol,3-bromo-2-hydroxyanisole,acmc-1ch0f,phenol,2-bromo-6-methoxy,2-bromanyl-6-methoxy-phenol,ksc560s4p
11019958
2-bromo-6-methoxyphenol
COC1=C(C(=CC=C1)Br)O
Applications
2-Bromo-6-methoxyphenol is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Quantity 1g
Melting Point 62°C to 65°C
Formula Weight 203.04
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-6-methoxyphenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning