Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-6-methyl-3-nitropyridine, 98%

Click to view available options
Quantity:
1g
5g
25g
374633-31-5
C6H5BrN2O2
217.022
MFCD03095212
IHHLJCDGOSZFTB-UHFFFAOYSA-N
2-bromo-3-nitro-6-methylpyridine,2-bromo-3-nitro-6-picoline,2-bromo-6-methyl-3-nitro-pyridine,2-bromo-3-nitro-6-methyl pyridine,pubchem5954,acmc-209it3,ksc496c9p,6-bromo-5-nitro-2-picoline,pyridine, 2-bromo-6-methyl-3-nitro,2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
24728013
2-bromo-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])Br
374633-31-5
C6H5BrN2O2
217.022
MFCD03095212
IHHLJCDGOSZFTB-UHFFFAOYSA-N
2-bromo-3-nitro-6-methylpyridine,2-bromo-3-nitro-6-picoline,2-bromo-6-methyl-3-nitro-pyridine,2-bromo-3-nitro-6-methyl pyridine,pubchem5954,acmc-209it3,ksc496c9p,6-bromo-5-nitro-2-picoline,pyridine, 2-bromo-6-methyl-3-nitro,2-bromo-6-methyl-3-nitropyridinemethyl 6-bromomethyl nicotinate
24728013
2-bromo-6-methyl-3-nitropyridine
CC1=NC(=C(C=C1)[N+](=O)[O-])Br
Applications
It is used as pharmaceutical intermediate.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 1g
Color White
Beilstein 138200
Formula Weight 217.02
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-methyl-3-nitropyridine

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning