Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-6-nitrophenol, 97%, Acros Organics™

Click to view available options
Quantity:
5g
25g
13073-25-1
C6H4BrNO3
218.01
VEJSIOPQKQXJAT-UHFFFAOYSA-N
2-bromo-6-nitrophenol,phenol, 2-bromo-6-nitro,2-bromo-6-nitro-phenol,6-bromo-2-nitrophenol,6-nitro-2-bromophenol,3-bromo-2-hydroxynitrobenzene,pubchem3114,2-brom-6-nitrophenol,2-nitro-6-bromophenol,acmc-209bjb
13545453
C1=CC(=C(C(=C1)Br)O)[N+](=O)[O-]
13073-25-1
C6H4BrNO3
218.01
VEJSIOPQKQXJAT-UHFFFAOYSA-N
2-bromo-6-nitrophenol,phenol, 2-bromo-6-nitro,2-bromo-6-nitro-phenol,6-bromo-2-nitrophenol,6-nitro-2-bromophenol,3-bromo-2-hydroxynitrobenzene,pubchem3114,2-brom-6-nitrophenol,2-nitro-6-bromophenol,acmc-209bjb
13545453
C1=CC(=C(C(=C1)Br)O)[N+](=O)[O-]
Melting Point 67.0°C to 69.0°C
Quantity 25g
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 218.01
Percent Purity 97%
Chemical Name or Material 2-Bromo-6-nitrophenol

GHS H Statement
Harmful if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Wear protective gloves.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning