Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-6-nitrotoluene, 98%

Click to view available options
Quantity:
1g
5g
25g
55289-35-5
C7H6BrNO2
216.034
MFCD00009792
LYTNSGFSAXWBCA-UHFFFAOYSA-N
2-bromo-6-nitrotoluene,6-bromo-2-nitrotoluene,3-bromo-2-methylnitrobenzene,1-bromo-2-methyl-3-nitro-benzene,benzene, 1-bromo-2-methyl-3-nitro,bromo 2--6-nitrotoluene,2-bromo-6-nitrotoluene,2-nitro-6-bromotoluene,3-bromo-2-methyl-1-nitrobenzene,1-brom-2-methyl-3-nitrobenzol,pubchem3823
123537
1-bromo-2-methyl-3-nitrobenzene
CC1=C(C=CC=C1Br)[N+](=O)[O-]
55289-35-5
C7H6BrNO2
216.034
MFCD00009792
LYTNSGFSAXWBCA-UHFFFAOYSA-N
2-bromo-6-nitrotoluene,6-bromo-2-nitrotoluene,3-bromo-2-methylnitrobenzene,1-bromo-2-methyl-3-nitro-benzene,benzene, 1-bromo-2-methyl-3-nitro,bromo 2--6-nitrotoluene,2-bromo-6-nitrotoluene,2-nitro-6-bromotoluene,3-bromo-2-methyl-1-nitrobenzene,1-brom-2-methyl-3-nitrobenzol,pubchem3823
123537
1-bromo-2-methyl-3-nitrobenzene
CC1=C(C=CC=C1Br)[N+](=O)[O-]
Quantity 25g
Flash Point >110°C (230°F)
Melting Point 37°C to 42°C
Boiling Point 108°C to 110°C (3.3mmHg)
Beilstein 2502581
Formula Weight 216.04
Percent Purity 98%
Chemical Name or Material 2-Bromo-6-nitrotoluene

GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning