Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-9-fluorenone, 95%, ACROS Organics™

Click to view available options
Quantity:
100mg
Packaging:
Glass bottle
3096-56-8
C13H7BrO
259.102
MFCD00010789
MTCARZDHUIEYMB-UHFFFAOYSA-N
2-bromo-9-fluorenone,2-bromo-9h-fluoren-9-one,9h-fluoren-9-one, 2-bromo,2-bromanylfluoren-9-one,acmc-209hjk,2-bromo-9h-fluorenone,ksc318o8f
725831
2-bromofluoren-9-one
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)Br
3096-56-8
C13H7BrO
259.102
MFCD00010789
MTCARZDHUIEYMB-UHFFFAOYSA-N
2-bromo-9-fluorenone,2-bromo-9h-fluoren-9-one,9h-fluoren-9-one, 2-bromo,2-bromanylfluoren-9-one,acmc-209hjk,2-bromo-9h-fluorenone,ksc318o8f
725831
2-bromofluoren-9-one
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)Br
Melting Point 143°C to 149°C
Quantity 100mg
Color Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 259.1
Physical Form Powder
Percent Purity ≥94%
Chemical Name or Material 2-Bromo-9-fluorenone, 95%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning