Alfa Aesar™ 2-Bromo-N-methylaniline, 95%

Manufacturer: Alfa Aesar™ H5516603

Quantity 1g

 View more versions of this product

Catalog No. AAH5516603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6832-87-7
    Formula Weight 186.05
    MDL Number MFCD06798064
    Chemical Name or Material 2-Bromo-N-methylaniline

    Specifications

    Boiling Point 107° to 109°C (12mmHg)
    Formula Weight 186.05
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Color Brown
    MDL Number MFCD06798064
    CAS 6832-87-7
    Physical Form Liquid
    Health Hazard 1 Warning
    Refractive Index 1.6070
    Density 1.589g/mL
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material 2-Bromo-N-methylaniline
    Assay 95%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit