Alfa Aesar™ 2-Bromo-N,N-dimethylaniline, 98+%

Manufacturer: Alfa Aesar™ B2556414

Quantity 25g

 View more versions of this product

Catalog No. AAB2556414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 698-00-0
    Molecular Formula C8H10BrN
    Formula Weight 200.08
    MDL Number MFCD00013522
    Beilstein 2205741
    Synonym N,N-Dimethyl-2-bromoaniline
    Chemical Name or Material 2-Bromo-N,N-dimethylaniline

    Specifications

    Formula Weight 200.08
    MDL Number MFCD00013522
    Boiling Point 108°C (14mmHg)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H10BrN
    CAS 698-00-0
    Beilstein 2205741
    Synonym N,N-Dimethyl-2-bromoaniline
    Quantity 25g
    Chemical Name or Material 2-Bromo-N,N-dimethylaniline
    Assay ≥98%

    GHS H Statement
    H301-H311-H332-H315-H319
    Toxic if swallowed.
    Toxic in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit