Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromo-N,N-dimethylaniline, 98+%

Click to view available options
Quantity:
5g
25g
698-00-0
C8H10BrN
200.079
MFCD00013522
ONMSBNJJCUCYED-UHFFFAOYSA-N
2-bromo-n,n-dimethylbenzenamine,n,n-dimethyl-2-bromoaniline,o-bromo-n,n-dimethylaniline,o-bromo-n,n'-dimethylaniline,benzenamine, bromo-n,n-dimethyl,pubchem3759,bromo-n,n-dimethylaniline,acmc-1az6p,2-bromophenyl dimethylamine,2-dimethylamino bromobenzene
136527
2-bromo-N,N-dimethylaniline
CN(C)C1=CC=CC=C1Br
698-00-0
C8H10BrN
200.079
MFCD00013522
ONMSBNJJCUCYED-UHFFFAOYSA-N
2-bromo-n,n-dimethylbenzenamine,n,n-dimethyl-2-bromoaniline,o-bromo-n,n-dimethylaniline,o-bromo-n,n'-dimethylaniline,benzenamine, bromo-n,n-dimethyl,pubchem3759,bromo-n,n-dimethylaniline,acmc-1az6p,2-bromophenyl dimethylamine,2-dimethylamino bromobenzene
136527
2-bromo-N,N-dimethylaniline
CN(C)C1=CC=CC=C1Br
Quantity 5g
Boiling Point 108°C (14mmHg)
Beilstein 2205741
Formula Weight 200.08
Percent Purity ≥98%
Chemical Name or Material 2-Bromo-N,N-dimethylaniline

GHS H Statement
H301-H311-H332-H315-H319
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger