Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2'-Bromoacetophenone, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2142-69-0
C8H7BrO
199.05
PIMNFNXBTGPCIL-UHFFFAOYSA-N
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
75060
1-(2-bromophenyl)ethanone
CC(=O)C1=CC=CC=C1Br
2142-69-0
C8H7BrO
199.05
PIMNFNXBTGPCIL-UHFFFAOYSA-N
2'-bromoacetophenone, 1-2-bromophenyl ethanone, o-bromoacetophenone, ethanone, 1-2-bromophenyl, 1-acetyl-2-bromobenzene, 1-2-bromophenyl ethan-1-one, acetophenone, 2'-bromo, 1-2-bromomphenyl ethanone, acetophenone, o-bromo, o-bromophenyl methyl ketone
75060
1-(2-bromophenyl)ethanone
CC(=O)C1=CC=CC=C1Br
Specific Gravity 1.476
Quantity 25g
Boiling Point 116.0°C to 117.0°C (12.0 mmHg)
Density 1.4760g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point >110°C
Refractive Index 1.5650 to 1.5690
Color Orange to Yellow
Assay Percent Range 98% min. (GC)
Linear Formula BrC6H4COCH3
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chlorofom,eth
Formula Weight 199.05
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material 2'-Bromoacetophenone
Show More Show Less

GHS Signal Word: Warning

EINECSNumber : 218-398-1