Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoacetophenone 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Plastic bottle
70-11-1
C8H7BrO
199.05
MFCD00000195
LIGACIXOYTUXAW-UHFFFAOYSA-N
2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo
6259
CHEBI:51846
2-bromo-1-phenylethanone
C1=CC=C(C=C1)C(=O)CBr
70-11-1
C8H7BrO
199.05
MFCD00000195
LIGACIXOYTUXAW-UHFFFAOYSA-N
2-bromoacetophenone,phenacyl bromide,2-bromo-1-phenylethan-1-one,bromoacetophenone,ethanone, 2-bromo-1-phenyl,bromomethyl phenyl ketone,alpha-bromoacetophenone,stauffer 4644,omega-bromoacetophenone,acetophenone, 2-bromo
6259
CHEBI:51846
2-bromo-1-phenylethanone
C1=CC=C(C=C1)C(=O)CBr
Acylation reagent
Quantity 2.5kg
Flash Point >112°C
Melting Point 46.0°C to 51.0°C
Boiling Point 135.0°C (18.0 mmHg)
Color Brown-Green to White
Infrared Spectrum Authentic
Packaging Plastic bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H5COCH2Br
Beilstein 07, 283
Fieser 02,316; 12,381
Merck Index 15, 1409
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: freely soluble in alcohol,benzene,chloroform,and ether
Formula Weight 199.05
Physical Form Crystals or Powder
Percent Purity 98%
Chemical Name or Material 2-Bromoacetophenone, 98%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-724-9

TSCA : TSCA