Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Bromoacetyl)-6-methoxynaphthalene, 96%, Acros Organics

Click to view available options
Quantity:
1g
5g
10262-65-4
C13H11BrO2
279.13
HHHKEQGAGUAOQI-UHFFFAOYSA-N
2-bromoacetyl-6-methoxynaphthalene,2-bromo-1-6-methoxy-2-naphthyl ethanone,2-bromo-6'-methoxy-2'-acetonaphthone,2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one,2-bromo-1-6-methoxynaphthalen-2-yl ethanone,ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl,enamine_005243,2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone
193452
2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr
10262-65-4
C13H11BrO2
279.13
HHHKEQGAGUAOQI-UHFFFAOYSA-N
2-bromoacetyl-6-methoxynaphthalene,2-bromo-1-6-methoxy-2-naphthyl ethanone,2-bromo-6'-methoxy-2'-acetonaphthone,2-bromo-1-6-methoxynaphthalen-2-yl ethan-1-one,2-bromo-1-6-methoxynaphthalen-2-yl ethanone,ethanone, 2-bromo-1-6-methoxy-2-naphthalenyl,enamine_005243,2-bromo-1-6-methoxy-naphthalen-2-yl-ethanone
193452
2-bromo-1-(6-methoxynaphthalen-2-yl)ethanone
COC1=CC2=C(C=C1)C=C(C=C2)C(=O)CBr
Quantity 5g
Melting Point 112.0°C
Assay Percent Range 95% min. (HPLC)
Solubility Information Solubility in water: insoluble
Formula Weight 279.13
Percent Purity 96%
Chemical Name or Material 2-(Bromoacetyl)-6-methoxynaphthalene

GHS Signal Word: Danger