Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-(Bromoacetyl)naphthalene, 98%

Click to view available options
Quantity:
10g
50g
250g
613-54-7
C12H9BrO
249.107
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-2'-acetonaphthone,bromomethyl 2-naphthyl ketone,2-bromoacetyl naphthalene,2-bromo-1-naphthalen-2-yl ethanone,2-bromo-1-2-naphthyl-1-ethanone,ethanone, 2-bromo-1-2-naphthalenyl,2-bromo-1-2-naphthyl ethanone,2-bromo-1-naphthalen-2-yl ethan-1-one,alpha-bromo-2'-acetonaphthone,2-bromo-1-naphthalen-2-yl-ethanone
69179
2-bromo-1-naphthalen-2-ylethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
613-54-7
C12H9BrO
249.107
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-2'-acetonaphthone,bromomethyl 2-naphthyl ketone,2-bromoacetyl naphthalene,2-bromo-1-naphthalen-2-yl ethanone,2-bromo-1-2-naphthyl-1-ethanone,ethanone, 2-bromo-1-2-naphthalenyl,2-bromo-1-2-naphthyl ethanone,2-bromo-1-naphthalen-2-yl ethan-1-one,alpha-bromo-2'-acetonaphthone,2-bromo-1-naphthalen-2-yl-ethanone
69179
2-bromo-1-naphthalen-2-ylethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
Melting Point 80°C to 85°C
Quantity 250g
UN Number UN3261
Beilstein 639153
Formula Weight 249.11
Percent Purity 98%
Chemical Name or Material 2-(Bromoacetyl)naphthalene

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.