Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2-Bromoadamantane, 98%
Click to view available options
Quantity:
5g
25g
Specifications
Quantity | 25g |
Melting Point | 136°C to 141°C |
Beilstein | 4655839 |
Formula Weight | 215.14 |
Percent Purity | 98% |
Chemical Name or Material | 2-Bromoadamantane |
Safety and Handling
EINECSNumber : 230-777-3
TSCA : No
Recommended Storage : Ambient temperatures