Alfa Aesar™ 2-Bromoadamantane, 98%

Manufacturer: Alfa Aesar™ B2276006

Quantity 5g

 View more versions of this product

Catalog No. AAB2276006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7314-85-4
    Molecular Formula C10H15Br
    Formula Weight 215.14
    MDL Number MFCD00074745
    Beilstein 4655839
    Synonym 2-Adamantyl bromide
    Chemical Name or Material 2-Bromoadamantane

    Specifications

    Formula Weight 215.14
    Molecular Formula C10H15Br
    MDL Number MFCD00074745
    Melting Point 136° to 141°C
    Quantity 5g
    CAS 7314-85-4
    Beilstein 4655839
    Synonym 2-Adamantyl bromide
    Chemical Name or Material 2-Bromoadamantane
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit