Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoanthracene, 98%, Acros Organics™

Click to view available options
Quantity:
1g
5g
7321-27-9
C14H9Br
257.13
PYXBCVWIECUMDW-UHFFFAOYSA-N
2-bromo-anthracene,anthracene, 2-bromo,pubchem19987,acmc-209opw,ksc377a5l
12346099
2-bromoanthracene
C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Br
7321-27-9
C14H9Br
257.13
PYXBCVWIECUMDW-UHFFFAOYSA-N
2-bromo-anthracene,anthracene, 2-bromo,pubchem19987,acmc-209opw,ksc377a5l
12346099
2-bromoanthracene
C1=CC=C2C=C3C=C(C=CC3=CC2=C1)Br
Quantity 5g
Infrared Spectrum Authentic
Melting Point 220.0°C to 221.5°C
Assay Percent Range 97.5% min. (GC)
Formula Weight 257.13
Percent Purity 98%
Chemical Name or Material 2-Bromoanthracene

GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning