Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromobenzaldoxime, 96%

Click to view available options
Quantity:
1g
5g
25g
34158-72-0
C7H6BrNO
200.035
MFCD00082734
PSIRFUPZHPEKAE-WEVVVXLNSA-N
2-bromobenzaldoxime,e-2-bromobenzaldehyde oxime,2-bromobenzaldehyde oxime,e-n-2-bromophenyl methylidene hydroxylamine,e-1-2-bromophenyl-n-hydroxymethanimine,z-n-2-bromophenyl methylidene hydroxylamine,2-bromobenzal-doxime,pubchem3731,2-bromophenyl hydroxyimino methane,ne-n-2-bromophenyl methylidene hydroxylamine
6876022
(NE)-N-[(2-bromophenyl)methylidene]hydroxylamine
C1=CC=C(C(=C1)C=NO)Br
34158-72-0
C7H6BrNO
200.035
MFCD00082734
PSIRFUPZHPEKAE-WEVVVXLNSA-N
2-bromobenzaldoxime,e-2-bromobenzaldehyde oxime,2-bromobenzaldehyde oxime,e-n-2-bromophenyl methylidene hydroxylamine,e-1-2-bromophenyl-n-hydroxymethanimine,z-n-2-bromophenyl methylidene hydroxylamine,2-bromobenzal-doxime,pubchem3731,2-bromophenyl hydroxyimino methane,ne-n-2-bromophenyl methylidene hydroxylamine
6876022
(NE)-N-[(2-bromophenyl)methylidene]hydroxylamine
C1=CC=C(C(=C1)C=NO)Br
Quantity 5g
Melting Point 100°C to 104°C
Beilstein 471599
Formula Weight 200.03
Percent Purity 96%
Chemical Name or Material 2-Bromobenzaldoxime

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning