Alfa Aesar™ 2-Bromobenzotrifluoride, 99%

Manufacturer: Alfa Aesar™ A1013422

Quantity 100g

 View more versions of this product

Catalog No. AAA1013422

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 392-83-6
    Molecular Formula C7H4BrF3
    Formula Weight 225.01
    MDL Number MFCD00000373
    Beilstein 1945750
    Synonym 2-(Trifluoromethyl)bromobenzene
    Chemical Name or Material 2-Bromobenzotrifluoride

    Specifications

    Formula Weight 225.01
    CAS 392-83-6
    Beilstein 1945750
    Quantity 100g
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    RTECS Number XS7980000
    TSCA TSCA
    MDL Number MFCD00000373
    Synonym 2-(Trifluoromethyl)bromobenzene
    Boiling Point 167° to 168°C
    Flash Point 68°C (154°F)
    Odor Aromatic
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Molecular Formula C7H4BrF3
    Chemical Name or Material 2-Bromobenzotrifluoride
    Assay 99%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit