Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzyl alcohol, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
25g
Packaging:
Glass bottle
18982-54-2
C7H7BrO
187.04
MFCD00004600
IOWGHQGLUMEZKG-UHFFFAOYSA-N
2-bromobenzyl alcohol,2-bromophenyl methanol,o-bromobenzyl alcohol,benzenemethanol, 2-bromo,o-bromo-benzyl alcohol,2-bromobenzylalcohol,2-bromophenyl methan-1-ol,o-bromobenzylalcohol,2-bromo-benzylalcohol,pubchem3746
72850
(2-bromophenyl)methanol
C1=CC=C(C(=C1)CO)Br
18982-54-2
C7H7BrO
187.04
MFCD00004600
IOWGHQGLUMEZKG-UHFFFAOYSA-N
2-bromobenzyl alcohol,2-bromophenyl methanol,o-bromobenzyl alcohol,benzenemethanol, 2-bromo,o-bromo-benzyl alcohol,2-bromobenzylalcohol,2-bromophenyl methan-1-ol,o-bromobenzylalcohol,2-bromo-benzylalcohol,pubchem3746
72850
(2-bromophenyl)methanol
C1=CC=C(C(=C1)CO)Br
Melting Point 78.0°C to 82.0°C
Quantity 25g
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4CH2OH
Beilstein 06, 445
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in most organic solvents
Formula Weight 187.04
Percent Purity 98%
Chemical Name or Material 2-Bromobenzyl alcohol, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Do not eat,drink or smoke when using this product.
Wash face,hands and any exposed skin thoroughly after handling.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Use only outdoors or in a well-ventilated area.

GHS Signal Word: Warning

EINECSNumber : 242-719-4

TSCA : TSCA