Alfa Aesar™ 2-Bromobenzyl bromide, 98%

Manufacturer: Alfa Aesar™ A1128530

Quantity 250g

 View more versions of this product

Catalog No. AAA1128530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3433-80-5
    Molecular Formula C7H6Br2
    Formula Weight 249.94
    MDL Number MFCD00000173
    UN Number UN1759
    Beilstein 971015
    Synonym alpha,2-Dibromotoluene
    Chemical Name or Material 2-Bromobenzyl bromide

    Specifications

    Formula Weight 249.94
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Quantity 250g
    UN Number UN1759
    Synonym alpha,2-Dibromotoluene
    Melting Point 29° to 32°C
    Flash Point >110°C (230°F)
    Molecular Formula C7H6Br2
    Beilstein 971015
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
    Boiling Point 127° to 129°C (19mmHg)
    CAS 3433-80-5
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    MDL Number MFCD00000173
    Chemical Name or Material 2-Bromobenzyl bromide
    Assay 98%

    Description

    Applications
    2-Bromobenzyl bromide was used in the synthesis of substituted quinazolines and 1,2,3,4-tetrahydroquinazolines, 2- and 3-substituted indenes, tris-2-bromotribenzylamine3.

    Solubility
    Insoluble in water.

    Notes
    Store away from strong bases and oxidizing agents. Incompatible with amines, alcohols, steel.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling
  • Promotions

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit