Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobenzylamine hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
5465-63-4
C7H8BrClN
221.50
MFCD00012853
ZANQNQMPTMEAIP-UHFFFAOYSA-N
2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
2724202
(2-bromophenyl)methanamine;hydrochloride
[Cl].NCC1=CC=CC=C1Br
5465-63-4
C7H8BrClN
221.50
MFCD00012853
ZANQNQMPTMEAIP-UHFFFAOYSA-N
2-bromobenzylamine hydrochloride, 2-bromophenyl methanamine hydrochloride, o-bromobenzylamine hydrochloride, 1-2-bromophenyl methanamine hydrochloride, 2-bromobenzylamine hcl, 1-2-bromophenyl methylamine hydrochloride, 2-bromophenyl methylamine, chloride, benzenemethanamine, 2-bromo-, hydrochloride, pubchem3749, acmc-1aup2
2724202
(2-bromophenyl)methanamine;hydrochloride
[Cl].NCC1=CC=CC=C1Br
Packaging Glass bottle
Melting Point 225°C to 230°C
Water 0.5% max. (K.F.)
Quantity 25g
Color White to Yellow
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (Argentometry)
Linear Formula BrC6H4CH2NH2·HCl
Beilstein 12, 1074
Formula Weight 222.52
Physical Form Crystalline Powder, Crystals and/or Chunks
Percent Purity 97%
Chemical Name or Material 2-Bromobenzylamine hydrochloride, 97%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA