Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromobiphenyl, 98%

Click to view available options
Quantity:
1g
5g
25g
2052-07-5
C12H9Br
233.11
MFCD00000065
KTADSLDAUJLZGL-UHFFFAOYSA-N
2-bromobiphenyl,o-bromobiphenyl,2-bromo-biphenyl,1,1'-biphenyl, 2-bromo,2-bromo-1,1'-biphenyl,biphenyl, 2-bromo,2-bromo-diphenyl,bromobiphenyl,2-bromodiphenyl,1,1'-biphenyl, bromo
16329
1-bromo-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2Br
2052-07-5
C12H9Br
233.11
MFCD00000065
KTADSLDAUJLZGL-UHFFFAOYSA-N
2-bromobiphenyl,o-bromobiphenyl,2-bromo-biphenyl,1,1'-biphenyl, 2-bromo,2-bromo-1,1'-biphenyl,biphenyl, 2-bromo,2-bromo-diphenyl,bromobiphenyl,2-bromodiphenyl,1,1'-biphenyl, bromo
16329
1-bromo-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2Br
Applications
It is used as an intermediate of organic synthesis and pharmaceutical. It also can react with norborn-2-ene to get 5-phenyl-1,2,3,4,4a,8b-hexahydro-1,4-methanobiphenylene, in synthesis of a novel trispirocyclic hydrocarbon. It is used as a reference compound to investigate extraction efficiency of solid-phase micro extraction fibers, based on methacrylic acid-trimethylolpropanetrimethacrylate copolymers.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Melting Point 0°C to 2°C
Flash Point >110°C (230°F)
Quantity 25g
Boiling Point 298°C
Beilstein 1863156
Formula Weight 233.12
Percent Purity 98%
Chemical Name or Material 2-Bromobiphenyl