Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromobiphenyl, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
2052-07-5
C12H9Br
233.11
MFCD00000065
KTADSLDAUJLZGL-UHFFFAOYSA-N
2-bromobiphenyl,o-bromobiphenyl,2-bromo-biphenyl,1,1'-biphenyl, 2-bromo,2-bromo-1,1'-biphenyl,biphenyl, 2-bromo,2-bromo-diphenyl,bromobiphenyl,2-bromodiphenyl,1,1'-biphenyl, bromo
16329
1-bromo-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2Br
2052-07-5
C12H9Br
233.11
MFCD00000065
KTADSLDAUJLZGL-UHFFFAOYSA-N
2-bromobiphenyl,o-bromobiphenyl,2-bromo-biphenyl,1,1'-biphenyl, 2-bromo,2-bromo-1,1'-biphenyl,biphenyl, 2-bromo,2-bromo-diphenyl,bromobiphenyl,2-bromodiphenyl,1,1'-biphenyl, bromo
16329
1-bromo-2-phenylbenzene
C1=CC=C(C=C1)C2=CC=CC=C2Br
Quantity 1g
Specific Gravity 1.352
Density 1.3520g/mL
Boiling Point 297.0°C to 298.0°C
Flash Point 113°C
Refractive Index 1.6280 to 1.6300
Melting Point 1.5°C to 2.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Linear Formula C6H5C6H4Br
Formula Weight 233.11
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 2-Bromobiphenyl
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-141-3