Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Bromodibenzofuran, 98%

Click to view available options
Quantity:
250mg
1g
5g
86-76-0
C12H7BrO
247.091
MFCD00092338
CRJISNQTZDMKQD-UHFFFAOYSA-N
2-bromodibenzo b,d furan,2-bromo-dibenzofuran,dibenzofuran, 2-bromo,dibenzofuran, 2-bromo-8ci 9ci,2-bromodibenzo b furan,2-bromo-dibenzofuran,4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene,4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene,4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene,4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
6856
2-bromodibenzofuran
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br
86-76-0
C12H7BrO
247.091
MFCD00092338
CRJISNQTZDMKQD-UHFFFAOYSA-N
2-bromodibenzo b,d furan,2-bromo-dibenzofuran,dibenzofuran, 2-bromo,dibenzofuran, 2-bromo-8ci 9ci,2-bromodibenzo b furan,2-bromo-dibenzofuran,4-bromo-8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6,10,12-hexaene,4-bromo-8-oxatricyclo 7.4.0.0 2 , trideca-1 9 ,2 7 ,3,5,10,12-hexaene,4-bromo-8-oxatricyclo 7.4.0.0?,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene,4-bromo-8-oxatricyclo 7.4.0.0^ 2,7 trideca-1 13 ,2,4,6,9,11-hexaene
6856
2-bromodibenzofuran
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)Br
Applications
2-Bromodibenzofuran is used as a reagent in Friedel-Crafts acylation reactions.

Solubility
Soluble in hot methanol.

Notes
Incompatible with oxidizing agents.
Melting Point 108°C to 111°C
Quantity 250mg
Beilstein 142199
Formula Weight 247.09
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 2-Bromodibenzofuran

GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P301+P312-P337+P313-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning