Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethyl ether, 90%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
5414-19-7
C4H8Br2O
231.92
FOZVXADQAHVUSV-UHFFFAOYSA-N
bis 2-bromoethyl ether,1-bromo-2-2-bromoethoxy ethane,2-bromoethyl ether,2,2'-dibromodiethyl ether,ethane, 1,1'-oxybis 2-bromo,ether, bis 2-bromoethyl,beta,beta'-dibromodiethyl ether,2,2-dibromodiethyl ether,2-bromoethylether
21521
1-bromo-2-(2-bromoethoxy)ethane
C(CBr)OCCBr
5414-19-7
C4H8Br2O
231.92
FOZVXADQAHVUSV-UHFFFAOYSA-N
bis 2-bromoethyl ether,1-bromo-2-2-bromoethoxy ethane,2-bromoethyl ether,2,2'-dibromodiethyl ether,ethane, 1,1'-oxybis 2-bromo,ether, bis 2-bromoethyl,beta,beta'-dibromodiethyl ether,2,2-dibromodiethyl ether,2-bromoethylether
21521
1-bromo-2-(2-bromoethoxy)ethane
C(CBr)OCCBr
Boiling Point 92°C to 93°C (12.0mmHg)
Flash Point 85°C
Packaging Glass Bottle
Specific Gravity 1.845
Density 1.8450g/mL
Quantity 5mL
Assay Percent Range 88% min. (GC)
Linear Formula [Br(CH2)2]2O
Formula Weight 231.92
Percent Purity 90%
Chemical Name or Material 2-Bromoethyl ether
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

EINECSNumber : 226-504-2