Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromoethyl ethyl ether, 90%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
592-55-2
C4H9BrO
153.02
MFCD00000237
MMYKTRPLXXWLBC-UHFFFAOYSA-N
2-bromoethyl ethyl ether,2-ethoxyethyl bromide,2-bromoethoxyethane,ethane, 1-bromo-2-ethoxy,2-bromodiethyl ether,1-ethoxy-2-bromoethane,1-bromo-2-ethoxy-ethane,ether, 2-bromoethyl ethyl,2-bromoethylethylether,1-bromo-3-oxapentane
61141
1-bromo-2-ethoxyethane
CCOCCBr
592-55-2
C4H9BrO
153.02
MFCD00000237
MMYKTRPLXXWLBC-UHFFFAOYSA-N
2-bromoethyl ethyl ether,2-ethoxyethyl bromide,2-bromoethoxyethane,ethane, 1-bromo-2-ethoxy,2-bromodiethyl ether,1-ethoxy-2-bromoethane,1-bromo-2-ethoxy-ethane,ether, 2-bromoethyl ethyl,2-bromoethylethylether,1-bromo-3-oxapentane
61141
1-bromo-2-ethoxyethane
CCOCCBr
Density 1.3570g/mL
Flash Point 21°C
Boiling Point 125°C to 127°C
Specific Gravity 1.357
Refractive Index 1.4435 to 1.4515
Quantity 100g
Packaging Glass bottle
Color Colorless to Brown
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula BrCH2CH2OC2H5
Beilstein 01,338
Solubility Information Solubility in water: slightly soluble in water. Other solubilities: miscible in ethanol,ethyl ether
Formula Weight 153.02
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 2-Bromoethyl ethyl ether
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Danger

EINECSNumber : 209-763-6