Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-Bromoethyl)trimethylammonium Bromide 98%, Thermo Scientific Chemicals

Catalog Number AC152350050
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
2758-06-7
C5H13Br2N
246.97
MFCD00011865
OINMNSFDYTYXEQ-UHFFFAOYSA-M
2758-06-7
C5H13Br2N
246.97
MFCD00011865
OINMNSFDYTYXEQ-UHFFFAOYSA-M

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 240°C to 244°C
Color Beige to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula BrCH2CH2N(CH3)3Br
Quantity 5 g
Synonym 2-bromoethyl trimethylammonium bromide, 2-bromo-n,n,n-trimethylethanaminium bromide, 2-bromoethyltrimethylammonium bromide, bromcholin, hypercyl, kathesin, btab, btab the plant regulator, bcb the plant regulator, unii-kh1i7ru18l
SMILES C[N+](C)(C)CCBr.[Br-]
IUPAC Name 2-bromoethyl(trimethyl)azanium;bromide
PubChem CID 17689
Formula Weight 246.97
Percent Purity ≥97.5%
Physical Form Chunks and/or Powder
Chemical Name or Material (2-Bromoethyl)trimethylammonium bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 220-419-4
RTECSNumber BO9481000
TSCA TSCA

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit